Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Metal TeenPrep™ Adapter for 12 x 15 mL Tube Holder on FastPrep-24

Catalog No. 116002546 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-24, FastPrep-96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-600-2546 FastPrep-24, FastPrep-96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-600-2546 Supplier MP Biomedicals™ Supplier No. 116002546
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metal TeenPrep™ adapter for 12 x 15 mL tube holder on FastPrep-24 is used to grind or homogenize most difficult samples. It can resist high temperatures up to 450°C.

  • Product Overview: This is the strongest FastPrep-24™ adapter, intended for the most difficult and long-lasting lysis applications. Due to its metal construction, it can be continuously run on a FastPrep™ indefinitely without the possibility of de-polymerization. This all-metal adapter can resist high temperatures up to 450°C and can be sterilized by pyrolysis. Furthermore, the specially designed claws prevent tubes from auto-rotation during lysis, thus allowing continuous tracking of tube labels and/or bar codes. The major use of this adapter is for lysing highly contagious samples and samples suspected of harboring prions. Due to its ability for high-temperature sterilization, this is the safest solution for CNS tissue lysis and other applications where prions or other disease-causing agents may be present. 15 mL tubes metal adapter for FastPrep-24™.
  • To secure this tube holder adapter on FastPrep 24s, it is recommended to use the Ratchet Nut v3.0, blue (116005510), sold separately.
  • Capacity: 12 tubes
TRUSTED_SUSTAINABILITY

Specifications

Product Type FastPrep-24 Metal Adapters
Content And Storage Room Temperature
For Use With (Application) Nucleic Acid Isolation and Purification, Sample Lysis and Preparation
For Use With (Equipment) FastPrep-24, FastPrep-96
Holds 12 x 15 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.