Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Avidin from Egg White

Catalog No. ICN15040725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15040725 25 mg
ICN15040701 1 mg
ICN15040705 5 mg
ICN15040710 10 mg
ICN15040780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN15040725 Supplier MP Biomedicals™ Supplier No. 0215040725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avidin is a basic glycoprotein consisting of four essentially identical subunits

Avidin was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.

  • CAS Number: 1405-69-2
  • EC Number: 215-783-6
  • MDL Number: MFCD00130572
  • Format: White Crystalline Powder
  • Specific Activity: Approx. 10 to 15 units/mg protein
  • Soluble in water or dilute aqueous buffer
  • Isoelectric point: 10

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Source Chicken
Content And Storage 0°C
Form Lyophilized
For Use With (Application) Molecular Biology
Product Type Avidin from Egg White
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.