Learn More
MP Biomedicals™ Baby Rabbit Complement, 3-4 week old
Catalog No. MP08642961 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
MP08642961 5 mL
1 options
Catalog No. MP08642961 Supplier MP Biomedicals™ Supplier No. 08642961
Add to Cart
Add to Cart
Complement obtained from a pool of baby rabbits, 3-4 weeks old.
  • The rabbits are of United States origin.
  • The complement is supplied as a liquid and does not contain any preservatives.
  • Store the complement at -20oC. Avoid Freeze/Thaw cycles.
TRUSTED_SUSTAINABILITY
Specifications
Type Immunoassay Reagent: Baby Rabbit Complement
Quantity 5 mL
WARNING: Cancer - www.P65Warnings.ca.gov