Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bio-Imager Pen

Catalog No. ICN17010001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN17010001 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN17010001 Supplier MP Biomedicals™ Supplier No. 0217010001

missing translation for 'validMainframeMsg'

Designed for recording data and positioning information on membranes prior to exposure to X-ray film

  • Contains a non-toxic, phosphorescent writing material which casts a greenish-white glow in the dark
  • Produces optimal images in approximately 10-12 hours of X-ray exposure
  • Saves time, reduces cost and eliminates isotopic material use
  • Recharged by light

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
Includes Phophorescent writing material
Format Pen
For Use With (Application) Molecular biology and biochemistry, for recording data and positioning information on membranes prior to exposure to X-ray film
Storage Requirements 15° to 30°C
Product Type Bio Imager Pen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.