Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Collagen, Sterile

Catalog No. ICN19349220 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19349220 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19349220 Supplier MP Biomedicals™ Supplier No. 0219349220
Only null left
Add to Cart
Add to Cart

Aqueous solution

Contains 0.3% pure bovine collagen buffered with 0.1% acetate. 95% type I collagen, 5% type III collagen. pH (20 °C): 3.2-3.8.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Contains 0.3% pure Bovine Collagen Buffered with 0.1% Acetate; 95% Type I Collagen, 5% Type III Collagen
Name Collagen from Bovine
pH Range 1.7 to 2.1
Quantity 20 mL
Source Bovine
Storage Requirements 4°C
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.