Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Fibrinogen

Catalog No. ICN820225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN820225 100g
ICN820224 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN820225 Supplier MP Biomedicals™ Supplier No. 08820225
Only null left
Add to Cart
Add to Cart

Suitable as a substrate for cell culture and in clotting assays

Prepared by extensive washing of cryoprecipitate followed by a buffer extraction to remove unwanted proteins and precipitation with ammonium sulfate. Lyophilized powder. Clottable protein >90%.

Physical Characteristics

  • Appearance: Off-white powder
  • Clottable Protein (anhydrous): ≥90%
  • Moisture: ≤5%
  • Reported: Total protein, pH
Certifications: ISO, cGMP

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Mammalian Cell Culture
Name Bovine Fibrinogen 90% Clottable from Bovine Plasma
pH Range 6.4
Quantity 100g
Source Bovine Plasma
Storage Requirements <-10°C
Form Lyophilized
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.