Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein, Hammerstein Grade

Catalog No. ICN10128901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10128901 1 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10128901 Supplier MP Biomedicals™ Supplier No. 0210128901
Only null left
Add to Cart
Add to Cart

Casein is the name for a family of related phosphoproteins

Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin.

  • CAS Number: 9000-71-9
  • EC Number: 232-555-1
  • MDL Number: MFCD00081481
  • Molecular Weight: 75,000 to 100,000
  • Format: white powder
  • Soluble in alkaline aqueous solutions; very sparingly soluble in water and non-polar organic solvents
  • Moisture content: 6±2%
  • pH: 5.5 (2% suspension)

Used in production and characterization of phage-display recombinant antibody against carrageenans.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 75000 to 100000
Name Casein
pH Range 5.5
Quantity 1 lb.
Storage Requirements 15° to 30°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.