Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CellMaxx™ Bovine Albumin, Low IgG, Stem Cell Culture

Catalog No. p-4341461 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219989710 10 g
MP219989725 25 g
ICN989750 50 g
MP219989780 100 g
MP219989790 500 g
MP219989791 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP219989710 Supplier MP Biomedicals™ Supplier No. 0219989710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation

Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells; IgG removed to very low levels. Lipid-rich formulation can be substituted for Albumax I™.

Physical characteristics:

  • Form: Off white to brown powder
  • Purity: ≥98% (w/w)
  • Total Protein: ≥96%
  • IgG 0.005% w/w
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Lipid content
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Mammalian Cell Culture, Western Blot
Formulation Lipid-rich
Molecular Weight (g/mol) 66
Name CellMaxx™ Bovine Albumin, Low IgG
pH Range 6.5 to 7.5
Purification Method Chromatography
Quantity 10 g
Source Bovine
Storage Requirements 4°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.