Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Chorionic Gonadotropin (hCG)

Catalog No. ICN19859110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19859110 10 mg
ICN19859180 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19859110 Supplier MP Biomedicals™ Supplier No. 0219859110
Add to Cart
Add to Cart

From human pregnancy urine

Lyophilized.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) ∽39500
Name Chorionic Gonadotropin from Human Pregnancy Urine
Quantity 10 mg
Source Human Pregnancy Urine
Storage Requirements -20°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.