Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CIRCLEGROW™ Sterile Solution

Catalog No. MP130001044 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP130001044 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP130001044 Supplier MP Biomedicals™ Supplier No. 113000104
Add to Cart
Add to Cart

Superior plasmid yield through enhanced cell growth

Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium. CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death during the log phase of growth.

  • Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed
  • Reduce hands-on work and preparation time
  • Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
  • Obtain high yields of plasmid DNA from smaller culture volumes, and confidently leave cultures overnight or over the weekend
  • Capsules are the most convenient form of this unique growth media
  • Pre-mixed CIRCLEGROW is also available in bulk powder, single-use pouches and capsules

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Circlegrow Medium
Certifications/Compliance Molecular Biology Certified
Content And Storage 15° to 30°C
Detects E.coli, Plasmid DNA
Form Solution
Media Type Circlegrow
Product Type Culture Medium
Sterility Sterile
Target Organism Class Bacteria
Test Type Growth Promotion Testing
Fill Volume 500 mL
Final Product Type Plasmid DNA
Formulation Proprietary formulation-Yeast Extract, Tryptone Broth, and Vitamins, Biological buffers
For Use With (Application) Cell Culture, Proteomics, Purification and Detection
pH 6.9
Product Line CIRCLEGROW
Quantity 500 mL
Specimen Type E. coli
Protein Source Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.