Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cell Lysis Solution for Tissues and Cells

Catalog No. 116540409 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
110 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-654-0409 110 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-654-0409 Supplier MP Biomedicals™ Supplier No. 116540409
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell Lysis/DNA solubilizing solution for tissues and cell extractions. Used in conjunction with a Protein Precipitation Solution (PPS). Component of the FastDNA Kit and FastDNA SPIN Kit.

  • CLS-TC is a colorless, precipitate-free solution, with a pH range of 5.8 to 6.2
  • This reagent is stable at room temperature
  • Shelf life can be extended by storing at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C
Format Kit
For Use With (Application) Nucleic Acid Isolation and Purification
pH 5.8 to 6.2
Product Type CLS-TC
Quantity 110 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.