Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagen from Calf Skin

Catalog No. ICN16008483 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16008483 250 mg
ICN16008450 50 mg
ICN16008480 100 mg
ICN16008410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN16008483 Supplier MP Biomedicals™ Supplier No. 0216008483
Add to Cart
Add to Cart

Soluble form

Suitable for gel formation, platelet aggregation and assay of collagenase by viscometry. Prepared by method of Gallop, P.M. and Seifter, S., Meth. in Enzymol., VI: 635 (1963).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5mg/mL
For Use With (Application) Gel Formation, Platelet Aggregation, Collagenase Assay
Molecular Weight (g/mol) ∽1000000
Name Collagen from Calf skin
pH Range 3.7
Quantity 250 mg
Source Calf Skin
Storage Requirements 4°C
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.