Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Complement C1q Protein

Catalog No. ICN19139101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19139101 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19139101 Supplier MP Biomedicals™ Supplier No. 0219139101
Only null left
Add to Cart
Add to Cart

From human serum

Purity: Functionally and biochemically pure by PAGE. Concentration: 1mg/mL. Supplied in PBS, 40% glycerol.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.1mg/mL
Formulation Supplied in PBS, 40% Glycerol
Name Human Complement C1q
Purification Method SDS-PAGE
Quantity 1mL
Source Human Serum
Storage Requirements -70°C
Cross Reactivity Human Serum
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.