Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Complement, Guinea Pig Purified, 5mL

Catalog No. ICN55854 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55854 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55854 Supplier MP Biomedicals™ Supplier No. 0855854
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use in hemolytic titrations and cytotoxicity assays, and as a serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Product Type Guinea Pig complement
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.