Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Concanavalin A, FITC-Conjugated

Catalog No. ICN15324505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15324505 5 mg
ICN15324501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15324505 Supplier MP Biomedicals™ Supplier No. 0215324505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Concentration 5μg/mL
Formulation 0.05M Tris, 0.15M NaCl, 0.004M CaCl2, pH 7 to 7.2, contains Sodium Azide as a preservative
Name Rabbit Concanavalin A, FITC Conjugated from Jack bean
pH Range 7 to 7.2
Purification Method Gel Filtration
Quantity 5 mg
Source Jack Bean
Storage Requirements 0°C
Conjugate FITC
Cross Reactivity Rabbit
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.