Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Conical Tubes

Catalog No. MP116930000 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Packaging Quantity:
5 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
MP116930000 5 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116930000 Supplier MP Biomedicals™ Supplier No. 116930000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For nucleic acid purification and isolation

  • 50 x 15 mL conical tubes packaged in a bag
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Product Type Conical Tube
Packaging Quantity 5 Pk.
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.