Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cycloheximide

Catalog No. ICN19452705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19452705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: Toxic substance. Extremely hazardous to health (can be lethal) if inhaled, swallowed, or in contact with the skin. If contact occurs, consult a physician immediately.
Catalog No. ICN19452705 Supplier MP Biomedicals™ Supplier No. 0219452705
Add to Cart
Add to Cart
Warning: Toxic substance. Extremely hazardous to health (can be lethal) if inhaled, swallowed, or in contact with the skin. If contact occurs, consult a physician immediately.

  • Inhibits peptide synthesis

Also known as Acti-Dione and Naramycin A, this antibiotic inhibits peptide synthesis and is active against many molds, yeasts, and phytopathogenic fungi. Historically used in bacteriological media to isolate or count bacteria in the presence of yeasts and molds; current applications include apoptosis, gene expression, glycogenolysis, gluconeogenesis, ureogenesis, steroidogenesis, and plant growth regulation research. Supplied in crystalline form.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Product Type Antibiotic
Quantity 5 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.