Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ D-(+)- GALACTOSE (POWDER) Anhydrous

Catalog No. MP114008012 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114008012 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114008012 Supplier MP Biomedicals™ Supplier No. 114008012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

White anhydrous powder with applications in the areas of yeast cell culture and gene expression

  • Purity: ≥98%
  • Physical Description: White Powder
  • Formula Weight: 180.16
  • Formula: C6H12O6
  • CAS #: 59-23-4
  • FTIR: Conforms to Standard
  • Melting Point: 160-170°C

  • Moisture: <1%
  • Optical Rotation (c=4, water) +77° ± 5°

Yeast Cell Culture; Gene Expression

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Quantity 227 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.