Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ D-(+)- RAFFINOSE PENTAHYDRATE (POWDER)

Catalog No. MP114061522 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114061522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114061522 Supplier MP Biomedicals™ Supplier No. 114061522
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystalline white, pure (≥98%) raffinose pentahydrate powder with applications in the areas of yeast cell culture and gene expression

  • Purity:≥98%
  • Physical Description: White Crystalline Powder
  • Formula: C18H32O165H2O
  • Formula Weight: 594.5
  • CAS #:17629-30-0
  • FTIR: Conforms to Standard
  • Melting Point: 76- 86°C
  • Optical Rotation (c=4, water): 101-107°
  • Loss on Drying: 13 - 17%
  • TLC: One Spot
  • Sulphated Ash (dry basis): 0.1%

Applications:

Yeast Cell Culture; Gene Expression

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.