Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Dairy Butter Diet, Pellet

Catalog No. ICN96039310 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96039310 10 kg
ICN96039350 50 kg
ICN96039320 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN96039310 Supplier MP Biomedicals™ Supplier No. 0296039310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Mice

An atherogenic diet for mice.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Components Butter 17.84%, Corn Oil 0.98%, Sucrose 48.33%, Casein, purified High Nitro gen 19.33%, Cholesterol, U.S.P. 1.23%, Sodium cholate 0.48%, Aphacel, Non-Nutritive Bulk 4.67%, AIN Mineral Mixture 4.79%, AIN Vitamin Mixture 0.97%, Choline Chloride 0.97%, DL-Methionine 0.29%, Vitamin E 0.12%
Format Pellet
For Use With (Application) Atherogenic diet for Mice
Quantity 10 kg
Storage Requirements 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.