Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Dairy Butter Diet, Pellet

Catalog No. p-3533142 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 kg
20 kg
50 kg

For Mice

An atherogenic diet for mice.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Components Butter 17.84%, Corn Oil 0.98%, Sucrose 48.33%, Casein, purified High Nitro gen 19.33%, Cholesterol, U.S.P. 1.23%, Sodium cholate 0.48%, Aphacel, Non-Nutritive Bulk 4.67%, AIN Mineral Mixture 4.79%, AIN Vitamin Mixture 0.97%, Choline Chloride 0.97%, DL-Methionine 0.29%, Vitamin E 0.12%
Format Pellet
For Use With (Application) Atherogenic diet for Mice
Quantity 10 kg
Storage Requirements 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.