Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ DEAE-Dextran, Chloride Form

Catalog No. ICN19513350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19513350 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19513350 Supplier MP Biomedicals™ Supplier No. 0219513350

missing translation for 'validMainframeMsg'

Chloride form

  • Prepared from Dextran MW 500,000.
  • Nitrogen Content: Approx. 3.2%

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Powder
Product Type DEAE-Dextran
pH 5 to 7
Quantity 50 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.