Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Dextri-Maltose Supplement

Catalog No. ICN96004801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96004801 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN96004801 Supplier MP Biomedicals™ Supplier No. 0296004801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maltose and dextrins obtained by enzymatic digestion of corn flour by barley malt.

  • Maltodextrin, a polymer chain composed of D-glucose units linked by α (1,4) bonds
  • Used in the development of encapsulation and non-encapsulation vehicles for use in the food industry
  • Used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity
  • As a sugar-matrix forming agent, maltodextrin is used in the development of orally disintegrating tablets (ODT)
  • Maltose and dextrins obtained by enzymatic digestion of corn flour by barley malt
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Course Powder
For Use With (Application) Maltodextrin, a Polymer chain composed of D-glucose units linked by a (1, 4) bonds, is used in the development of encapsulation and nanencapsulation vehicles for use in the food industry, Maltodextrin is used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity
Quantity 1 kg
Storage Requirements 15°C to 30°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.