Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Egg White

Catalog No. ICN90163391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90163391 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN90163391 Supplier MP Biomedicals™ Supplier No. 0290163391

missing translation for 'validMainframeMsg'

Spray Dried

TRUSTED_SUSTAINABILITY

Specifications

Product Type Egg White
Format Powder
For Use With (Application) Egg whites have been used in studies of enzyme production of bioactive peptides that show strong antioxidant activity
Particle Shape Egg Solids
Quantity 1000 g
Storage Requirements 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.