Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ EY Tellurite Enrichment

Catalog No. ICN1004149 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1004149 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1004149 Supplier MP Biomedicals™ Supplier No. 091004149
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a defoamer component. In hydrated form, it is used in toothpaste as a hard abrasive to remove tooth plaque. SiO

  • For use with Baird-Parker Agar Base for isolating S. aureus
  • Key application: Bacteria Cell Culture
  • Concentration: 30% egg yolk suspended with 0.15% potassium tellurite
  • Detects: S. aureus (for isolation)
  • Format: Solution
  • Product Type: Enrichment, Emulsion
  • Target Organism Class: Bacteria (S. aureus)
  • Test Type: Isolation (for S. aureus)
TRUSTED_SUSTAINABILITY

Specifications

Appearance Light Yellow Solution
Color Light Yellow
Concentration 30% Egg Yolk Suspended with 0.15% Potassium Tellurite
Content And Storage 4°C
Additive Potassium Tellurite
Detects S. aureus
Form Liquid
Format Solution
Manufacturing Origin United States
Media Type EY Tellurite Enrichment
Target Organism Class Bacteria
Fill Volume 100 mL
Formulation Consists of 30% Egg Yolk suspended with 0.15% Potassium Tellurite
For Use With (Application) Used with Baird Parker Agar base for the isolation of S. aureus
Incubation Conditions 50°C for Preparation
No. per Pack 1
Quantity 100 mL
Transfer Volume 50 mL
Protein Source Egg Yolk
Source Egg Yolk
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.