Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ F-10 Nutrient Mixture (HAM), Powder

Catalog No. MP091040122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Color:
Pale Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
MP091040122 Pale Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091040122 Supplier MP Biomedicals™ Supplier No. 091040122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A nutrient mixture designed to cultivate a wide variety of mammalian and hybridoma cells. F-10 Nutrient Mixture (HAM) with L-Glutamine, without Sodium Bicarbonate.

  • Originally developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells, and mouse L-cells.
  • F-10 is formulated for use with or without serum, depending on the type of cells being cultured.
  • Demonstrated satisfactory growth of human diploid cells, white blood cells for chromosomal analysis, as well as primary rat, rabbit, and chicken tissue explants.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Nutrient Medium
Sterility Non-sterile
With Additives Glutamine
Without Additives No Sodium Bicarbonate
pH 7 to 8
Tested For Endotoxin, Mycoplasma
Color Pale Orange
Content And Storage 0° to 5°C
Description Makes 10 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.