Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ham's F-12 (1X) with L-glutamine

Catalog No. MP091242354 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091242354 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091242354 Supplier MP Biomedicals™ Supplier No. 091242354
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Well suited for the growth of primary rat hepatocytes and prostate epithelial cells.

Ham's F-12 medium was designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells. Ham's F-12 is well suited for the growth of primary rat hepatocytes and prostate epithelial cells. It is also the medium of choice for toxicity assays using CHO cells.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type F-12 Nutrient Mixture (HAM)
Sterility Sterile
Quantity 500 mL
pH 7.2 to 7.4
Content And Storage 0 to 5°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.