Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TeenPrep™ Adapter for 12 x 15 mL Tube Holder on FastPrep-24
GSA_VA

Catalog No. ICN6002526 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Holds:
12 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
ICN6002526 12 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN6002526 Supplier MP Biomedicals™ Supplier No. 116002526
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TeenPrep™ Adapter is a medium-size adapter that holds 12 x 15 mL Lysing Matrix tubes that allows frozen samples to be immediately processed with minimal hands-on manipulation.

Medium-size adapter that holds 12 x 15 mL Lysing Matrix tubes. It allows frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes. A wide range of disposable 15 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.

  • Application Notes: TeenPrep™ 12 X 15 mL Adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications.
  • Capacity: 12 tubes
  • Sample Volume: Up to 24 g

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Large Sample Volume Adapter
Sample Size 24 g
Content And Storage 15°C to 30°C
For Use With (Application) DNA and RNA isolation, Enzyme isolation and Protein production, natural products isolation, food preparation for quality analysis
Holds 12 x 15 mL
Weight (English) 1.115 lb.
Weight (Metric) 506 g
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.