Learn More
MP Biomedicals™ HiPrep™ Adapter for 48 x 2 mL Tube Holder on FastPrep-24
Shop All MP Biomedicals, Inc Products

Description
Double decker adapter that holds 48 x 2 mL Lysing Matrix tubes. The ergonomic design of the adapter ensures easy loading of Lysing Matrix tubes that remain securely sealed during the homogenization. It stands stable on the bench top and is commonly used as a tube rack. Ideally suited for high-throughput applications, up 48 samples can be homogenized simultaneously. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapters and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Application Notes: The FastPrep™-24 Instrument is ideally suited for high-throughput applications.
- Capacity: 48 tubes
- Sample Volume: Up to 10 g
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Sample Size | 10 g |
| Content And Storage | 15°C to 30°C |
| For Use With (Application) | High-throughput applications, up to 24 and 48 samples can be homogenized simultaneously |
| Holds | 48 x 2 mL |
| Weight (English) | 1.029 lb. |
| Weight (Metric) | 467 g |
| Product Type | High-throughput Adapter |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.