Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CoolPrep™ Adapter for 24 x 2 mL Tube Holder on FastPrep-24
GSA_VA

Catalog No. ICN6002528 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Holds:
24 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
ICN6002528 24 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN6002528 Supplier MP Biomedicals™ Supplier No. 116002528
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CoolPrep™ Adapter for FastPrep-24 instrument is a cryogenic adapter that allows simultaneous lysis of 24 x 2 mL samples. It is also used for extraction of sensitive biological compounds.

The CoolPrep 24 x 2 mL adapter is a cryogenic adapter which allows simultaneous lysis of 24 x 2 mL samples. Based on passive temperature control technology, the CoolPrep adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep™ precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. Most samples can be processed in 40 seconds or less.

  • Application: Ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
  • Capacity: 20 tubes
  • Sample Volume: Up to 5 g

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cryogenic Adapter
Sample Size 5 g
Content And Storage 15°C to 30°C
For Use With (Application) Extractions of any temperature-unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates and Enzymes from even the hardest samples to lyse
Holds 24 x 2 mL
Weight (English) 1.499 lb.
Weight (Metric) 680 g
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.