Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CoolTeenPrep™ Adapter for 6 x 15 mL Tube Holder on FastPrep-24
GSA_VA

Catalog No. ICN6002530 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Holds:
6 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
ICN6002530 6 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN6002530 Supplier MP Biomedicals™ Supplier No. 116002530
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CoolTeenPrep™ Adapter for 6 x 15 mL Tube Holder on FastPrep-24 for cryogenic grinding and lysis on FastPrep-24.

CoolTeenPrep™ 6 X 15 mL Adapter is a cryogenic adapter which allows simultaneous lysis of 6 x 15 mL samples. Based on passive temperature control technology, the CoolTeenPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray, is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep™ precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. A wide range of disposable 15 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.

  • Application: CoolTeenPrep™ 6 X 15 mL Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
  • Capacity: 15 tubes
  • Sample Volume: Up to 12 g

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cryogenic Adapter
Sample Size 12 g
Content And Storage 15°C to 30°C
For Use With (Application) Extractions of any temperature-unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates and Enzymes from even the hardest samples to lyse
Holds 6 x 15 mL
Weight (English) 1.382 lb.
Weight (Metric) 627 g

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.