Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastDNA™-96 Fecal DNA Extraction Kit

Catalog No. MP119696400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP119696400 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119696400 Supplier MP Biomedicals™ Supplier No. 119696400

missing translation for 'validMainframeMsg'

FastDNA™-96 Fecal DNA Extraction Kit is designed to isolate bacterial, protist, or host DNA from up to 80 mg of wet or dry stool, and even from caecum samples, through a purification process that eliminates PCR inhibitors.

  • Product Overview: The FastDNA-96™ Fecal DNA Kit isolates bacterial, protist, or host DNA from up to 80 mg of wet or dry stool, and even from caecum samples, through a purification process that eliminates PCR inhibitors. Fecal microbes, fungi, parasites, and other stool organisms are first lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
  • Applications: The FastDNA-96™ Fecal DNA Kit quickly and efficiently isolates inhibitor-free, PCR-ready genomic DNA from stool samples (i.e. humans, primates, birds, rodents, cattle, canine, feline, porcine, etc.).
  • Sample Type: Mammalian Stool (i.e. humans, primates, birds, rodents, cattle, canine, feline, porcine, etc.)
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Sample Type Mammalian Stool
No. of Reactions 2 x 96 Preps
Product Type FastDNA-96 Kits
Quantity 1 Ea.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.