Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastDNA™-96 Fungal and Bacterial DNA Kit, 2 x 96 Preps

Catalog No. MP119696300 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP119696300 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119696300 Supplier MP Biomedicals™ Supplier No. 119696300

missing translation for 'validMainframeMsg'

FastDNA™-96 Fungal and Bacterial DNA Kit is used for rapid, high-throughput isolation of PCR-quality DNA from tough-to-lyse fungi and bacteria.

  • Product Overview: The FastDNA™-96 Fungal and Bacterial DNA Kit is designed for the simple and rapid isolation of, PCR-quality DNA from tough-to-lyse fungi and bacteria in approximately 40 minutes. Gram(+/-) bacteria, fungi, including A. fumigatus, C. albicans, N. crassa, S. cerevisiae, S.pombe, and mycelium, as well as spores, nematodes, pollen, and even mammalian cells are easily lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
  • Applications: FastDNA™-96 Fungal/Bacterial DNA Kit is used for rapid isolation of PCR-quality DNA from tough-to-lyse fungi and bacteria in approximately 40 minutes.
  • Sample Type: Spores, Bacteria (Gram +/-), Fungi, Yeast, Algae
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Sample Type Spores, Bacteria (Gram +/-), Fungi, Yeast, Algae
No. of Reactions 2 x 96 Preps
Product Type FastDNA-96 Kits
Quantity 1 Ea.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.