Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastDNA™-96 Plant/Seed DNA Kit

Catalog No. mp119696600 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP119696600 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119696600 Supplier MP Biomedicals™ Supplier No. 119696600

FastDNA™-96 Plant/Seed DNA Kit is used for rapid, high-throughput isolation of PCR-ready genomic DNA from plants tissues and seeds. For use with the FastPrep-96™ Instrument.

  • Product Overview: Rapid, High-Throughput Isolation of PCR-Ready Genomic DNA from Plant and Seed Samples The FastDNA™-96 Plant and Seed DNA Kit is designed for the simple and rapid isolation of PCR-quality DNA from vegetative samples in approximately 50 minutes. Plant samples including stems, roots, leaves, buds, flowers, fruits and seeds are efficiently lysed in approximately 60 seconds with the FastPrep-96™ Instrument. Up to 80 mg samples are placed into each 1.2 mL tube of a FastDNA-96™ Lysing Matrix Rack (96-deep well plate) containing 2.0 mm specially stabilized Zirconium oxide lysing particles, which are designed to efficiently lyse a wide variety of plant specimens, while in the presence of a specially formulated lysis solution. The cleared lysate is then applied to a binding matrix to isolate the DNA, and spin column plates are used to simplify liquid handling. Eluted DNA is then applied to the MP-96 Inhibitor Removal Plate which removes tannins, polyphenols and polysaccharides that can inhibit PCR. Purified DNA is eluted in an EDTA-free, DNA elution solution, and is ready for downstream applications including digestion, electrophoresis, arrays and PCR (A260/A280 ratios ≥1.8). Yield is typically 5 μg of total DNA eluted in 50 to 100 μL of elution solution. This procedure can be performed in minutes, and there is no need for organic denaturants or proteinases.
  • Applications: The FastDNA™-96 Plant/Seed DNA Kit for FastPrep™-96 is used for the simple, rapid isolation of inhibitor-free, PCR-quality DNA from a variety of plant sample sources including leaves, stems, buds, flowers, fruit, and seeds. Eluted DNA can be used for downstream molecular-based applications including PCR, arrays, digestion, electrophoresis etc.
  • Sample Source: Plant Tissues
  • Sample Type: Seeds, Plants, Roots, Buds, Flowers, Fruits, Leaves, Stems
  • Sample Volume: Up to 80 mg
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Kit
Sample Type Seeds, Plants, Roots, Buds, Flowers, Fruits, Leaves, Stems
No. of Reactions 2 x 96 Preps
Product Type FastDNA-96 Kits
Quantity 1 Ea.
Sample Volume Up to 80 mg
Sample Source Plant Tissues

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.