Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastPROTEIN™ BlueMatrix

Catalog No. p-4426020 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 Preps

Requires

FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0 mL tubes, Microcentrifuge tubes (2.0 mL and 1.5 mL), 1x PBS or Desired Lysis Buffer

FastPROTEIN™ BlueMatrix Kit designed for the rapid extraction of proteins from gram (+/-) bacteria in a few seconds.

  • Kit or Assay Type: Protein Isolation, Mini-prep
  • Molecule to be Enriched: Proteins
  • Sample Type: Bacteria, Gram +/-
  • Tube Size: 2 mL
  • Matrix Composition: Matrix B, 0.1 mm zirconium silicate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blue matrix kit
Quantity 50 Preps
Includes Matrix B, 0.1 mm Zirconium Silicate, Tubes
Content And Storage 15°C to 30°C
Capacity 2 mL
For Use With (Application) Rapid isolation and purification of Protein from bacterial cultures
Matrix FastProtein Blue Matrix
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.