Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastRNA™ SPIN Kit for Microbes

Catalog No. MP116020050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116020050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Research Use Only
Catalog No. MP116020050 Supplier MP Biomedicals™ Supplier No. 116020050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Research Use Only

FastRNA™ SPIN Kit for Microbes isolates total RNA from bacterial cell cultures and other microbial sources in approximately 15 minutes. Does not use phenol or reducing agents.

  • Product Overview: The FastRNA™ SPIN Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures and other microbial sources in approximately 15 minutes using a zirconium silicate Lysing Matrix (0.1 mm) to lyse bacteria and SPIN columns for the purification process.
  • Sample Source: Microorganisms
  • Sample Type: Bacteria (Gram +/-)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
For Use With (Application) Nucleic Acid Isolation and Purification
Includes Lysing Matrix B (0.1 mm Zirconium Silicate), 50 x 2 mL Tubes, 50 mL FastRNA Lysis Buffer, 25 mL FastRNA Prep Buffer, 12 mL FastRNA Wash Buffer, 1 mL Water (DNase/RNase-free), 50 Tubes FastRNA Spin Column 1, 50 Tubes FastRNA Spin Column 2, 100 x 2 mL Collection Tubes
No. of Reactions 50 Preps
Product Type FastRNA-24 Kits
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.