Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fat Diet, High, Modified, Paste

Catalog No. 0296019210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-960-19210 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-960-19210 Supplier MP Biomedicals™ Supplier No. 0296019210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fat Diet, High, Modified in paste form.

  • Not available in pellet form
  • Normal High Fat Diet (ICN96019210) modified to contain essential trace minerals, methionine and fiber
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Paste
Quantity 10 kg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.