Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold

Catalog No. p-91791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 100 mL
Color:
Golden Brown
Sterilization Method:
Filtration
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Sterilization Method
09-291-6749 1 x 100 mL Golden Brown Filtration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-291-6749 Supplier MP Biomedicals™ Supplier No. 092916749
Only null left
Add to Cart
Add to Cart

Superior for hybridoma cloning

  • Collected from a USDA regulated and monitored abattoir by technicians who work “onsite”
  • Aseptically collected via cardiac puncture
  • Supplied sterile, with no detectable mycoplasma or viral infection
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USDA
Color Golden Brown
Content And Storage -20°C
Form Liquid
Product Type Fetal Bovine Serum
Quantity 1 x 100 mL
Concentration 3.1 to 4.4 g/dL
Sterility Sterile
Sterilization Method Filtration
Endotoxin Concentration <1 ng/mL
For Use With (Application) Used for cell culture and hybridoma cloning
Hemoglobin Concentration 27 mg/dL
Mycoplasma Detected No
Osmolality 280 to 325 mOsm/kg
pH 6.7 to 7.8
USDA Safety Testing Included USDA Inspected
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.