Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fetal Bovine Serum, CellECT™ Silver

Catalog No. ICN2916154 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2916154 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2916154 Supplier MP Biomedicals™ Supplier No. 092916154/DEL

Aseptically collected via cardiac puncture

•    Collected from a USDA regulated and monitored abbatoir by technicians who work onsite
•    Careful handling minimizes cell lysis and other detrimental factors, thereby reducing possible lysozyme and hemoglobin contamination
•    Supplied sterile, with no detectable mycoplasma or viral infection
•    Endotoxin level: slightly more than 1 ng/mL

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USDA
Color Golden Brown
Content And Storage -20°C
Form Liquid
Product Type Fetal Bovine Serum
Quantity 1 x 500 mL
Concentration 3.1 to 4.4g/dL
Sterility Sterile
Sterilization Method Filtration
Endotoxin Concentration Slightly exceed 1ng/mL
For Use With (Application) Used to supplement cell culture media for the optimal growth of many cell types, effective in promoting and sustaining growth of vertebrate mammalian and insect cells
Hemoglobin Concentration 27mg/dL
Mycoplasma Detected No
Osmolality 280 to 325mOsm/kg
pH 6.7 to 7.8
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.