Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fibrinogen from Bovine Plasma

Catalog No. ICN15112201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15112201 1 g
ICN15112205 5 g
ICN15112210 10 g
ICN15112225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15112201 Supplier MP Biomedicals™ Supplier No. 0215112201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Off-white shiny flaky powder

Fibrinogen is blood protein that is involved in clotting and is converted to fibrin by thrombin. It consists of three non-identical pairs of disulfidebonded chains. The a-chain has an approximate molecular weight of 63.5 kDa, the b-chain 56 kDa, and the g-chain 47 kDa.

  • Molecular weight: 340kDa

Cell Biology and Analysis, Molecular biology

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Biology and Analysis, Molecular Biology
Molecular Weight (g/mol) 340
Name Fibrinogen from Bovine Plasma
Quantity 1 g
Source Bovine Plasma
Storage Requirements 0°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.