Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fibronectin, ≥90%

Catalog No. ICN15002501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15002501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15002501 Supplier MP Biomedicals™ Supplier No. 0215002501
Only null left
Add to Cart
Add to Cart

Derived from Bovine Plasma

Adhesive glycoproteins and are found both as cell surface proteins and in plasma. They are large glycoproteins found in cells and extracellular fluids. They are involved in a wide range of biological activities including cell adhesion to extracellular matrices, blood coagulation and wound healing

  • Format: White lyophilized powder
  • Molecular weight: 220,000 (Lit.)
  • Soluble in water

Dentistry; Cell Biology and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 to 50μg/mL
For Use With (Application) Cell Biology and Analysis
Molecular Weight (g/mol) 220000
Name Fibronectin from Bovine Plasma
Quantity 1 mg
Source Bovine Plasma
Storage Requirements 4°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.