Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fibronectin from Bovine Plasma

Catalog No. ICN15002505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15002505 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15002505 Supplier MP Biomedicals™ Supplier No. 0215002505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Involved in cell adhesion to extracellular matrices, blood coagulation and wound healing

  • Format: White fiberous material
  • Purity: ≥90%
  • Molecular weight: 220,000d
  • Sterile filtered

Dentistry; Cell Biology and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 to 50μg/mL
For Use With (Application) Cell Biology and Analysis
Molecular Weight (g/mol) 220000
Name Fibronectin from Bovine Plasma
Quantity 5 mg
Source Bovine Plasma
Storage Requirements 4°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.