Learn More
MP Biomedicals™ Fibronectin, Containing <1μg of Factor VIII and Fibrinogen
Shop All MP Biomedicals, Inc ProductsDescription
Used as a substrate for the promotion of attachment and replication of cells in culture.
Specifications
Specifications
| For Use With (Application) | Cell Attachment and Proliferation |
| Molecular Weight (g/mol) | 440000 |
| Name | Fibronectin from Human Plasma |
| pH Range | 7.3 |
| Preparation Method | Cohn fraction I of Human Plasma by a modification of the procedure |
| Quantity | 500 μg |
| Source | Human Plasma |
| Storage Requirements | 0°C |
| Form | Lyophilized |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.