Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Flaxseed Oil

Catalog No. ICN96043201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
450 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96043201 450 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN96043201 Supplier MP Biomedicals™ Supplier No. 0296043201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supplement for Animal Diet also know as Linseed Oil

  • CAS Number 8001-26-1
  • Refractive Index: n20/D 1.4795 (lit.)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Liquid
Quantity 450 g
Storage Requirements 15°C to 30°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.