Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FLUCONAZOLE

Catalog No. 0219898650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
02-198-98650 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-198-98650 Supplier MP Biomedicals™ Supplier No. 0219898650
Add to Cart
Add to Cart

Selective inhibitor of fungal cytochrome P-450 sterol C-14 α-demethylation

A subclass of the synthetic triazole antifungal agents

  • Physical Description: White crystalline powder
  • Molecular Formula: C13H12F2N6O
  • CAS Number: 86386-73-4
  • Synonyms: 2,4-Difluoro-1-bis(1H-1,2,4-triazol-1-ylmethyl) benzyl alcohol; 2,4-Difluoro-α,α1- bis(1H-1,2,4-triazol-1-ylmethyl) benzyl alcohol; α-(2,4-Difluorophenyl)-α-(1H-1,2,4-triazol-1-yl methyl)-1H-1,2,4-triazole-1-ethanol
  • Molecular Weight: 306.27
  • Solubility: Soluble in methanol and chloroform; sparingly soluble in acetone; very slightly to insoluble in water or saline

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.