Learn More
Description
- Geneclean™ EZ-Glassmilk™ is supplied with GeneClean™ III Kit which will bind up to radioactive isotope-labeled ssDNA or dsDNA that is purified from agarose thus resulting in purified DNA for further use.
- Geneclean™ EZ-Glassmilk™ allows flexibility of reaction size and volume
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Glass Milk |
| For Use With (Application) | Nucleic Acid Isolation & Purification |
| Product Type | Geneclean™ Slurry |
| Quantity | 3 mL |
Safety and Handling
Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.