Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Geneclean™ Glassmilk™ Silica Matrix

Catalog No. 111001404 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1404 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1404 Supplier MP Biomedicals™ Supplier No. 111001404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Geneclean™ Glassmilk™ Silica Matrix allows flexibility of reaction size and volume. Glassmilk™ slurry is a specially prepared aqeuous suspension of propietary silica matrix.

  • Geneclean™ Glassmilk™ is supplied with GeneClean™ Kit, which will bind to radioactive isotope-labeled ssDNA or dsDNA that is purified from agarose thus resulting in purified DNA for further use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 593-84-0
Content And Storage Room Temperature
Format Kit
Isolation Technology Silica Matrix Technology
For Use With (Application) Nucleic acid purification
Product Type Glassmilk™ Slurry
Quantity 1.5 mL
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.