Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Geneclean™ Spin Kit 200 bp to 300 kb

Catalog No. 111101200 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
100 Preps
300 Preps
50 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-110-1200 50 Preps
11-110-1400 100 Preps
11-110-1600 300 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-110-1200 Supplier MP Biomedicals™ Supplier No. 111101200
Add to Cart
Add to Cart

Used to purify fragments of DNA from 200 bp to 300 kbp from solutions and agarose gels. The GENECLEAN™ SPIN kit uses SPIN filters, which prevent silica particle carry-over into cleaned DNA. Glassmilk™ slurry allows flexibility of reaction size and volume.

  • The GENECLEAN™ SPIN kit simplifies the process of purifying DNA into three easy steps: Bind, Wash and Elute
  • Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications
  • Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA
TRUSTED_SUSTAINABILITY

Specifications

CAS 593-84-0
Format Kit
Sample Type Enzymatic Reactions, Solutions, Agarose Gels
For Use With (Application) Isolate DNA from TAE or TBE buffered agarose gels and solutions
Includes SPIN filters
No. of Reactions 50 Preps
pH Geneclean SPIN New Wash pH Range: 7.4 to 7.7, GC Spin Elution Solution pH Range: 6.8 to 7.2
Product Type Spin Kit
Sample Volume Up to ∼300 mg Gel Slice or up to ∼300 μL DNA Solution
Synonym DNA Clean-Up Kit
Sample Source Gels and Solutions
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.