Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit for PCR

Catalog No. MP111103400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
100 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP111103400 100 Preps
MP111103200 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP111103400 Supplier MP Biomedicals™ Supplier No. 111103400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Geneclean™ Turbo Kit optimized for PCR purification of DNA fragments of sizes 100 bp to 10 kb.

  • Ready for direct use
  • Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities
  • Applications: DNA Clean-up
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Glass Milk
Sample Type Post amplification PCR reaction mixture (oil-free)
For Use With (Application) Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples
Includes GeneClean™ Turbo™ Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution
No. of Reactions 100 Preps
Product Type DNA Purification Kit
Sample Volume Up to 100 μL PCR Reaction Solution
Sample Source Gels and Solutions

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.