Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit, 100 Preps

Catalog No. MP111102400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
100 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP111102400 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP111102400 Supplier MP Biomedicals™ Supplier No. 111102400
Add to Cart
Add to Cart

For nucleic acid isolation and purification

GeneClean™ Turbo Kit is a flexible kit designed for purification of DNA fragments of sizes 0.1 kb to 300 kb from TAE or TBE buffered agarose gels, PCR reactions, and other enzymatic solutions. It purifies larger fragments due to less manipulation of bound DNA. Process with either vacuum or centrifugation & save time with patented Turbo filter design.

Application Notes

GENECLEAN™ Turbo Kit is used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions, and other enzymatic solutions. Purified DNA can be used in enzymatic reactions, conventional or automated sequencing, cloning, and microarray analysis.

Usage Statement

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Key Applications

Purification of DNA fragments from agarose gels; PCR reactions | Elimination of proteins from enzymatic reactions

TRUSTED_SUSTAINABILITY

Specifications

Format Geneclean Imbedded Turbo Spin Column
Isolation Technology Glass Milk
For Use With (Application) Nucleic Acid isolation and purification
Includes 110mL Geneclean™ Turbo™ Salt solution, 11mL Geneclean™ Turbo™ wash concentrate, 100 Geneclean™ Turbo™ cartridges, 100 Geneclean™
No. of Reactions 100 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.